Assay Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106694.1 | NP_001100164.1 | 1 - 2 | 419 | 101 | |
GenBank mRNA | BC169047.1 | - | 1 - 2 | 413 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
