Assay Details
Gene Symbol: Tnfrsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025773.1 | NP_001020944.1 | 1 - 2 | 404 | 62 | |
GenBank mRNA | BC097483.1 | - | 1 - 2 | 404 | 62 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097483.1, NM_001025773.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
