Assay Details
Gene Symbol: Hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025409.1 | NP_001020580.1 | 2 - 3 | 187 | 83 | |
GenBank mRNA | BC059156.1 | - | 2 - 3 | 204 | 83 | |
BC097943.1 | - | 2 - 3 | 187 | 83 | ||
BC107476.1 | - | 2 - 3 | 184 | 83 |
Gene Symbol: Hdac1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053446.1 | NP_445898.1 | 1 | 169 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
