Assay Details
Gene Symbol: Hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025409.1 | NP_001020580.1 | 3 - 4 | 303 | 90 | |
GenBank mRNA | BC059156.1 | - | 3 - 4 | 320 | 90 | |
BC097943.1 | - | 3 - 4 | 303 | 90 | ||
BC107476.1 | - | 3 - 4 | 300 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
