Assay Details
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022383.2 | NP_071778.2 | 7 - 8 | 744 | 83 | |
XM_006238830.2 | XP_006238892.1 | 7 - 8 | 844 | 83 | ||
XM_006238831.2 | XP_006238893.1 | 7 - 8 | 841 | 83 | ||
XM_006238832.2 | XP_006238894.1 | 7 - 8 | 822 | 83 | ||
GenBank mRNA | BC089801.1 | - | 7 - 8 | 744 | 83 | |
L11930.1 | - | 6 - 7 | 646 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
