Assay Details
Gene Symbol: Wasf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013167.1 | NP_001013185.1 | 2 - 3 | 329 | 61 | |
XM_008764138.1 | XP_008762360.1 | 3 - 4 | 510 | 61 | ||
GenBank mRNA | BC089207.1 | - | 2 - 3 | 329 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
