Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239078.2 | XP_006239140.1 | 6 - 7 | 822 | 81 | |
XM_006239079.2 | XP_006239141.1 | 7 - 8 | 854 | 81 | ||
XM_006239080.2 | XP_006239142.1 | 7 - 8 | 854 | 81 | ||
XM_006239081.2 | XP_006239143.1 | 7 - 8 | 935 | 81 | ||
XM_006239082.2 | XP_006239144.1 | 6 - 7 | 866 | 81 | ||
GenBank mRNA | X57018.1 | - | 7 - 8 | 863 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
