Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024145.2 | NP_077059.2 | 1 - 2 | 188 | 94 | |
XM_006239081.2 | XP_006239143.1 | 1 - 2 | 205 | 93 | ||
GenBank mRNA | BC062025.1 | - | 1 - 2 | 188 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC062025.1, NM_024145.2, XM_006239081.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
