Assay Details
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013935.1 | NP_001013957.1 | 5 - 6 | 570 | 104 | |
XM_006239124.2 | XP_006239186.1 | 5 - 6 | 866 | 104 | ||
XM_006239125.2 | XP_006239187.1 | 5 - 6 | 644 | 104 | ||
GenBank mRNA | BC083692.1 | - | 5 - 6 | 570 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
