Assay Details
Gene Symbol: Extl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107985.1 | NP_001101455.1 | 10 - 11 | 1893 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P625531 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P161818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60816 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P625531 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161818 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P625531 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161818 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P625531 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P161818 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P368306 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32290 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63573 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10880513 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880511 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880515 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880505 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880512 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880514 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880506 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880510 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880507 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880508 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10880504 | - | - |
Rat Genome 230 2.0 Array | 1377472_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377472_at | - | - |
Rat Genome 230 2.0 Array | 1385365_at | - | - |
Rat Genome 230 2.0 Array | 1385365_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500028598.rn.1 | TC0500003813.rn.1 | RTA TC |
Rat Expression Set 230 | 1377472_at | - | - |
Rat Expression Set 230 | 1377472_PM_at | - | - |
Rat Expression Set 230 | 1385365_PM_at | - | - |
Rat Expression Set 230 | 1385365_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880504 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22124796 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354413 | - | - |
