Assay Details
Gene Symbol: Syf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC081735.1 | - | 1 - 2 | 28 | 102 | |
BC097289.1 | - | 1 - 2 | 53 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097289.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
