Assay Details
Gene Symbol: Syf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133417.1 | NP_596908.1 | 3 - 4 | 282 | 74 | |
GenBank mRNA | AF366369.1 | - | 3 - 4 | 282 | 74 | |
BC081735.1 | - | 3 - 4 | 265 | 74 | ||
BC097289.1 | - | 3 - 4 | 290 | 74 | ||
FQ234066.1 | - | 1 - 2 | 109 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
