Assay Details
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025050.1 | NP_001020221.1 | 7 - 8 | 741 | 101 | |
XM_008764064.1 | XP_008762286.1 | 8 - 9 | 741 | 101 | ||
GenBank mRNA | BC079274.1 | - | 7 - 8 | 741 | 101 | |
BC079293.1 | - | 8 - 9 | 720 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
