Assay Details
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025050.1 | NP_001020221.1 | 8 - 9 | 862 | 59 | |
XM_008764064.1 | XP_008762286.1 | 9 - 10 | 862 | 59 | ||
GenBank mRNA | BC079274.1 | - | 8 - 9 | 862 | 59 | |
BC079293.1 | - | 9 - 10 | 841 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
