Assay Details
Gene Symbol: Fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007554.1 | NP_001007555.1 | 4 - 5 | 547 | 93 | |
XM_008764273.1 | XP_008762495.1 | 4 - 5 | 1314 | 93 | ||
XM_008764274.1 | XP_008762496.1 | 3 - 4 | 477 | 93 | ||
XM_008764275.1 | XP_008762497.1 | 4 - 5 | 546 | 93 | ||
XM_008764276.1 | XP_008762498.1 | 4 - 5 | 697 | 93 | ||
GenBank mRNA | BC086398.1 | - | 4 - 5 | 547 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
