Assay Details
Gene Symbol: Fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764273.1 | XP_008762495.1 | 8 - 9 | 1896 | 72 | |
XM_008764274.1 | XP_008762496.1 | 7 - 8 | 1059 | 72 | ||
XM_008764275.1 | XP_008762497.1 | 8 - 9 | 1128 | 72 | ||
XM_008764276.1 | XP_008762498.1 | 8 - 9 | 1279 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
