Assay Details
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021582.1 | NP_067593.1 | 3 - 4 | 298 | 82 | |
GenBank mRNA | BC079180.1 | - | 3 - 4 | 298 | 82 | |
X98490.1 | - | 2 - 3 | 204 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
