Assay Details
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021582.1 | NP_067593.1 | 6 - 7 | 610 | 60 | |
GenBank mRNA | BC079180.1 | - | 6 - 7 | 610 | 60 | |
FQ228766.1 | - | 3 - 4 | 239 | 60 | ||
X98490.1 | - | 5 - 6 | 516 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
