Assay Details
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021582.1 | NP_067593.1 | 7 - 8 | 721 | 56 | |
GenBank mRNA | BC079180.1 | - | 7 - 8 | 721 | 56 | |
FQ228766.1 | - | 4 - 5 | 350 | 56 | ||
X98490.1 | - | 6 - 7 | 627 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
