Assay Details
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013241.1 | NP_001013259.1 | 5 - 6 | 715 | 78 | |
XM_006239434.2 | XP_006239496.1 | 5 - 6 | 857 | 78 | ||
XM_006239435.1 | XP_006239497.1 | 5 - 6 | 459 | 78 | ||
GenBank mRNA | BC087098.1 | - | 5 - 6 | 715 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
