Assay Details
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025737.1 | NP_001020908.1 | 2 - 3 | 378 | 106 | |
GenBank mRNA | BC097983.1 | - | 2 - 3 | 378 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
