Assay Details
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014193.2 | NP_001014215.2 | 4 - 5 | 989 | 70 | |
GenBank mRNA | BC085888.1 | - | 3 - 4 | 2311 | 70 | |
BF282923.1 | - | 1 - 2 | 70 | |||
FQ230600.1 | - | 4 - 5 | 974 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
