Assay Details
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017069.3 | NP_058765.3 | 7 - 8 | 1007 | 108 | |
XM_006229511.2 | XP_006229573.1 | 7 - 8 | 1119 | 108 | ||
GenBank mRNA | CB725341.1 | - | 1 - 2 | 43 | 108 | |
X51991.1 | - | 7 - 8 | 1014 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
