Assay Details
Gene Symbol: Nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009399.1 | NP_001009399.1 | 2 - 3 | 190 | 66 | |
XM_006229515.2 | XP_006229577.1 | 2 - 3 | 205 | 66 | ||
GenBank mRNA | BC087626.1 | - | 2 - 3 | 190 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
