Assay Details
Gene Symbol: Nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009399.1 | NP_001009399.1 | 6 - 7 | 772 | 82 | |
XM_006229515.2 | XP_006229577.1 | 6 - 7 | 787 | 82 | ||
GenBank mRNA | BC087626.1 | - | 6 - 7 | 772 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
