Assay Details
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004250.2 | NP_001004250.1 | 12 - 13 | 1380 | 92 | |
GenBank mRNA | BC078923.1 | - | 12 - 13 | 1382 | 92 | |
FQ210217.1 | - | 5 - 6 | 499 | 92 | ||
FQ213124.1 | - | 12 - 13 | 1391 | 92 | ||
FQ215592.1 | - | 8 - 9 | 864 | 92 | ||
FQ216746.1 | - | 12 - 13 | 1384 | 92 | ||
FQ224157.1 | - | 12 - 13 | 1388 | 92 | ||
FQ229052.1 | - | 12 - 13 | 1407 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
