Assay Details
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004250.2 | NP_001004250.1 | 7 - 8 | 826 | 88 | |
GenBank mRNA | BC078923.1 | - | 7 - 8 | 828 | 88 | |
FQ215592.1 | - | 3 - 4 | 310 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
