Assay Details
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199099.2 | NP_954530.1 | 3 - 4 | 300 | 62 | |
XM_006243751.2 | XP_006243813.1 | 3 - 4 | 341 | 62 | ||
GenBank mRNA | BC060585.1 | - | 3 - 4 | 301 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
