Assay Details
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106868.1 | NP_001100338.1 | 4 - 5 | 452 | 82 | |
XM_006244090.2 | XP_006244152.1 | 4 - 5 | 1354 | 82 | ||
XM_006244091.2 | XP_006244153.1 | 4 - 5 | 533 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
