Assay Details
Gene Symbol: Twf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135238.1 | NP_001128710.1 | 2 - 3 | 248 | 106 | |
GenBank mRNA | BC158614.1 | - | 2 - 3 | 196 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
