Assay Details
Gene Symbol: Sh3glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011929.1 | NP_001011929.1 | 4 - 5 | 788 | 133 | |
XM_006233431.2 | XP_006233493.1 | 4 - 5 | 803 | 133 | ||
XM_006233433.2 | XP_006233495.1 | 4 - 5 | 801 | 133 | ||
XM_006233435.2 | XP_006233497.1 | 4 - 5 | 800 | 133 | ||
GenBank mRNA | BC079085.1 | - | 4 - 5 | 788 | 133 | |
FQ234013.1 | - | 4 - 5 | 511 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
