Assay Details
Gene Symbol: Arhgef38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761544.1 | XP_008759766.1 | 250 | 76 |
Gene Symbol: LOC102556447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233345.2 | XP_006233407.1 | 2 - 3 | 602 | 76 | |
XM_008761533.1 | XP_008759755.1 | 2 - 3 | 602 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
