Assay Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025018.1 | NP_001020189.1 | 6 - 7 | 683 | 95 | |
XM_006233120.2 | XP_006233182.1 | 6 - 7 | 627 | 95 | ||
XM_006233121.2 | XP_006233183.1 | 5 - 6 | 567 | 95 | ||
XM_006233122.2 | XP_006233184.1 | 5 - 6 | 668 | 95 | ||
XM_006233123.2 | XP_006233185.1 | 6 - 7 | 629 | 95 | ||
GenBank mRNA | BC091251.1 | - | 6 - 7 | 683 | 95 | |
FQ218349.1 | - | 6 - 7 | 692 | 95 | ||
FQ228782.1 | - | 6 - 7 | 780 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
