Assay Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025018.1 | NP_001020189.1 | 1 - 2 | 141 | 59 | |
GenBank mRNA | BC091251.1 | - | 1 - 2 | 141 | 59 | |
FQ218349.1 | - | 1 - 2 | 150 | 59 | ||
FQ228782.1 | - | 1 - 2 | 238 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091251.1, NM_001025018.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
