Assay Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134549.2 | NP_001128021.2 | 3 - 4 | 1843 | 101 | |
XM_006233489.2 | XP_006233551.1 | 5 - 6 | 2127 | 101 | ||
XM_006233490.2 | XP_006233552.1 | 6 - 7 | 2717 | 101 | ||
XM_008761551.1 | XP_008759773.1 | 6 - 7 | 2156 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
