Assay Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134549.2 | NP_001128021.2 | 8 - 9 | 2379 | 152 | |
XM_006233489.2 | XP_006233551.1 | 10 - 11 | 2663 | 152 | ||
XM_006233490.2 | XP_006233552.1 | 11 - 12 | 3253 | 152 | ||
XM_006233491.2 | XP_006233553.1 | 10 - 11 | 2660 | 152 | ||
XM_008761551.1 | XP_008759773.1 | 11 - 12 | 2692 | 152 | ||
GenBank mRNA | FN806367.1 | - | 5 - 6 | 542 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
