Assay Details
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126120.1 | NP_001119592.1 | 6 - 7 | 838 | 74 | |
GenBank mRNA | BC083724.1 | - | 6 - 7 | 838 | 74 | |
FQ212950.1 | - | 6 - 7 | 894 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
