Assay Details
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085353.1 | NP_001078822.1 | 5 - 6 | 977 | 85 | |
NM_199408.2 | NP_955440.1 | 5 - 6 | 983 | 85 | ||
GenBank mRNA | AY569010.1 | - | 5 - 6 | 983 | 85 | |
BC062391.1 | - | 5 - 6 | 977 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
