Assay Details
Gene Symbol: Pitx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042505.1 | NP_001035970.1 | 1 - 2 | 550 | 66 | |
GenBank mRNA | AJ222971.1 | - | 1 - 2 | 558 | 66 | |
EF519321.1 | - | 1 - 2 | 40 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
