Assay Details
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106470.1 | NP_001099940.1 | 10 - 11 | 1164 | 122 | |
XM_006233266.2 | XP_006233328.1 | 10 - 11 | 1224 | 122 | ||
GenBank mRNA | BC166789.1 | - | 10 - 11 | 1070 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
