Assay Details
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106470.1 | NP_001099940.1 | 7 - 8 | 744 | 75 | |
XM_006233266.2 | XP_006233328.1 | 7 - 8 | 804 | 75 | ||
GenBank mRNA | BC166789.1 | - | 7 - 8 | 650 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
