Assay Details
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014042.1 | NP_001014064.1 | 5 - 6 | 852 | 57 | |
XM_006233150.2 | XP_006233212.1 | 5 - 6 | 1218 | 57 | ||
GenBank mRNA | BC087113.1 | - | 5 - 6 | 852 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
