Assay Details
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012183.2 | NP_001012183.1 | 3 - 4 | 340 | 67 | |
XM_006233520.2 | XP_006233582.1 | 4 - 5 | 576 | 67 | ||
GenBank mRNA | BC078927.1 | - | 3 - 4 | 342 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
