Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106471.1 | NP_001099941.1 | 10 - 11 | 1670 | 72 | |
GenBank mRNA | BC166793.1 | - | 10 - 11 | 1743 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P016500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73514 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P016500 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P016500 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P016500 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P532458 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59559 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10819071 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819070 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819067 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819073 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819072 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10819066 | - | - |
Rat Genome 230 2.0 Array | 1371850_at | - | - |
Rat Genome 230 2.0 Array | 1371850_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371850_at | - | - |
Affymetrix HT Rat Focus Array | 1371850_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200015318.rn.1 | TC0200002222.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200015319.rn.1 | TC0200002222.rn.1 | RTA TC |
Rat Expression Set 230 | 1371850_at | - | - |
Rat Expression Set 230 | 1371850_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10819066 | - | - |
Rat Genome U34 Set | rc_AA892829_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21846144 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372628 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350749 | - | - |
