Assay Details
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100518.1 | NP_001093988.1 | 4 - 5 | 871 | 82 | |
XM_008761495.1 | XP_008759717.1 | 4 - 5 | 927 | 82 | ||
GenBank mRNA | AY724508.1 | - | 4 - 5 | 871 | 82 | |
BC091291.1 | - | 2 - 3 | 227 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
