Assay Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031041.1 | NP_112303.1 | 3 - 4 | 274 | 141 | |
GenBank mRNA | BC085345.1 | - | 3 - 4 | 279 | 141 | |
FQ229769.1 | - | 3 - 4 | 262 | 141 | ||
FQ231337.1 | - | 3 - 4 | 304 | 141 | ||
X65948.1 | - | 3 - 4 | 274 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
