Assay Details
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015037.1 | NP_001015037.1 | 1387 | 77 | ||
GenBank mRNA | AY955395.1 | - | 1387 | 77 | ||
FQ218600.1 | - | 1125 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
