Assay Details
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033902.2 | NP_001029074.2 | 10 - 11 | 1273 | 91 | |
XM_008761469.1 | XP_008759691.1 | 8 - 9 | 1148 | 91 | ||
GenBank mRNA | BC103656.1 | - | 10 - 11 | 1206 | 91 | |
BC166503.1 | - | 10 - 11 | 1273 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
