Assay Details
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017282.1 | NP_058978.1 | 6 - 7 | 535 | 64 | |
GenBank mRNA | BC060575.1 | - | 6 - 7 | 556 | 64 | |
D10756.1 | - | 6 - 7 | 535 | 64 | ||
FQ218317.1 | - | 6 - 7 | 552 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
