Assay Details
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100794.1 | NP_001094264.1 | 7 - 8 | 906 | 93 | |
GenBank mRNA | BC086515.1 | - | 7 - 8 | 875 | 93 | |
BC166888.1 | - | 7 - 8 | 903 | 93 | ||
BC166920.1 | - | 7 - 8 | 903 | 93 | ||
FQ212650.1 | - | 7 - 8 | 885 | 93 | ||
FQ220191.1 | - | 7 - 8 | 885 | 93 | ||
FQ226064.1 | - | 7 - 8 | 892 | 93 | ||
FQ227364.1 | - | 7 - 8 | 892 | 93 | ||
FQ234387.1 | - | 7 - 8 | 892 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
